Learn More
Amsbio LLC Collagenase NB 4 Standard Grade (1g)
Supplier:  Amsbio LLC S1745401
Encompass_Preferred
Collagenase NB 4 Standard Grade is a natural enzyme mixture that contains collagenolytic and additional enzymatic activities including clostripain and neutral protease. The balanced ratio of these activities ensures gentle and efficient tissue dissociation. As Collagenase NB 4 Standard Grade has comparable enzymatic properties to Collagenase NB 5 Sterile Grade and Collagenase NB 6 GMP Grade, this enzyme may also be used in order to establish applications which have to meet sterile or GMP requirements like tissue engineering and transplantation.
Catalog No. 50-204-1257
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.