Learn More
Amsbio LLC Collagenase NB 4G Proved Grade (500 mg)
Supplier:  Amsbio LLC S1746501
Encompass
CollagenaseNB 4G Proved Grade is a crude collagenase which contains collagenolytic and additional enzymatic activities including clostripain and neutral protease. The balanced ratio of these activities leads to gentle and efficient tissue dissociation.
Catalog No. 50-204-1259
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.