Learn More
Amsbio LLC Collagenase NB 6 GMP Grade (1g)
Supplier:  Amsbio LLC N0002779
Encompass_Preferred
CollagenaseNB 6 GMP Grade is manufactured according to GMP guidelines. In addition, it is sterile according to European Pharmacopoeia. TSE safety of the manufacturing process was certified by the EDQM. A virus validation study and stability studies according to ICH guidelines were performed. Each lot possesses a low endotoxin level and is tested for abnormal toxicity according to Ph. Eur. CollagenaseNB 6 GMP Grade is a crude collagenase that contains collagenolytic and additional enzymatic activities including clostripain and neutral protease. The balanced ratio of these activities ensures gentle and efficient tissue dissociation.
Catalog No. 50-204-1264
Please contact customer service for price and further ordering instructions at 1-800-766-7000 or 1-800-926-1166.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.