Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Collagenase Type I from Clostridium histolyticum

Catalog No. MP21951095 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21951095 500 mg
MP219510901 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP21951095 Supplier MP Biomedicals™ Supplier No. 02195109.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For the preparation of isolated cell suspensions

The collagenase product is a mixture of enzymes secreted by C. histolyticum, with different products differentiated by the relative ratios of the 10-18 components found in the secreted enzymes. The main components are two collagenases, clostripain, and a neutral protease. The synergistic action of these enzymes degrade collagen and other intracellular material. The action of both collagenase enzymes and the neutral protease is necessary for effective release of cells from tissue. Various types of collagen are the natural substrates for collagenase.

Unit Definition: One unit will liberate peptides from collagen equivalent in ninhydrin color to 1.0μmol of L-leucine in 5 hr. at pH 7.4 and 37°C in presence of calcium ions. Contains clostripain, neutral protease and trypsin activities.

TRUSTED_SUSTAINABILITY

Specifications

Concentration >125 U/mg
Color Brown
For Use With (Application) Preparation of isolated cell suspensions
Content And Storage Store at 4°C. Desiccate.
Quantity 500 mg
Product Type Collagenase Type I
Form Lyophilized
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.