Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Collagenase Type I from Clostridium histolyticum

Catalog No. p-8015242 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
500 mg
1 g

For the preparation of isolated cell suspensions

The collagenase product is a mixture of enzymes secreted by C. histolyticum, with different products differentiated by the relative ratios of the 10-18 components found in the secreted enzymes. The main components are two collagenases, clostripain, and a neutral protease. The synergistic action of these enzymes degrade collagen and other intracellular material. The action of both collagenase enzymes and the neutral protease is necessary for effective release of cells from tissue. Various types of collagen are the natural substrates for collagenase.

Unit Definition: One unit will liberate peptides from collagen equivalent in ninhydrin color to 1.0μmol of L-leucine in 5 hr. at pH 7.4 and 37°C in presence of calcium ions. Contains clostripain, neutral protease and trypsin activities.

TRUSTED_SUSTAINABILITY

Specifications

Concentration >125 U/mg
Color Brown
For Use With (Application) Preparation of isolated cell suspensions
Content And Storage Store at 4°C. Desiccate.
Quantity 500 mg
Product Type Collagenase Type I
Form Lyophilized
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.