Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Collection Tubes for Vivacon™ 500, Bulk Pack (100 pcs)

Catalog No. 14556111 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Vivacon 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14-556-111 Vivacon 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-556-111 Supplier Sartorius Supplier No. VNCT01
Add to Cart
Add to Cart

Bulk pack (100 pieces) of collection reservoirs for use with Vivacon 500 centrifugal units.

Vivacon™ 500 are centrifugal units for fast DNA concentration and buffer exchange of 500μl samples. They are also the product of choice for protein fractionation applications. The use of Hydrosart™ membranes results in high recoveries from dilute samples. Complete recovery of the concentrate is guaranteed by a reverse spinning step.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Filter Type Ultrafiltration
For Use With (Equipment) Vivacon 500
Sterility Non-sterile
Includes 100 reservoirs
Type Tubing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.