Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Collibri™ PS DNA Library Prep Kits for Illumina Systems

Catalog No. A38614096W
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
24 Preps
96 Preps
Product Type:
PS DNA Library Prep Core Kit
PS DNA Library Prep Kit with CD Indexes
PS DNA Library Prep Kit with UD Indexes (Set A, 1-24)
PS DNA Library Prep Kit with UD Indexes (Set A-D, 1-96)
PS DNA Library Prep Kit with UD Indexes (Set B, 25-48)
PS DNA Library Prep Kit with UD Indexes (Set C, 49-72)
PS DNA Library Prep Kit with UD Indexes (Set D, 73-96)
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions Product Type
A38614096W 96 Preps PS DNA Library Prep Core Kit
A38612024W 24 Preps PS DNA Library Prep Core Kit
A38612024 24 Preps PS DNA Library Prep Kit with CD Indexes
A38614096 96 Preps PS DNA Library Prep Kit with CD Indexes
A38613024 24 Preps PS DNA Library Prep Kit with UD Indexes (Set A, 1-24)
A43611024 24 Preps PS DNA Library Prep Kit with UD Indexes (Set B, 25-48)
A43612024 24 Preps PS DNA Library Prep Kit with UD Indexes (Set C, 49-72)
A43613024 24 Preps PS DNA Library Prep Kit with UD Indexes (Set D, 73-96)
A38614196 96 Preps PS DNA Library Prep Kit with UD Indexes (Set A-D, 1-96)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. A38614096W Supplier Invitrogen™ Supplier No. A38614096W
Add to Cart
Add to Cart

The Invitrogen Collibri PS DNA Library Preparation kits are designed for next-generation sequencing (NGS) on Illumina systems. The kit supports whole-genome sequencing (WGS) applications and is suitable for all genomes, including FFPE samples.

The Collibri PS DNA Library Preparation Kit offers higher confidence in SNP and INDEL calling due to better coverage of challenging genomic regions from a wide range of inputs (1 ng to 1 μg) of physically sheared double-stranded DNA, cfDNA, or amplicons (≤300 bp) of any species. The entire library preparation protocol takes about three hours with a PCR workflow. Additionally, a PCR-free version is available for inputs starting at 500 ng DNA.

The Collibri PS DNA Library Preparation Kit offers higher confidence in SNP and INDEL calling due to better coverage of challenging genomic regions from a wide range of inputs (1 ng to 1 μg) of physically sheared double-stranded DNA, cfDNA, or amplicons (≤300 bp) of any species. The entire library preparation protocol takes about three hours with a PCR workflow. Additionally, a PCR-free version is available for inputs starting at 500 ng DNA.

The Collibri PS DNA Library Preparation Kit:
• Includes all necessary components in a single kit* for quick use
• Is empowered on miniaturization liquid handlers to cut on costs
• Provides in-process visual feedback for easier handling
• Is suitable for conventional automation to increase throughput

* The kit consists of enzymatic modification, size selection/clean-up, and indexed adapter-containing modules, except for the Core kit, which enables Collibri combination with desired indexed adapters.

The Collibri PS DNA Library Preparation Kit contains inert dyes that provide visual feedback of preparation progress: the reaction mixes change color in response to the addition of critical components at each step. The kit generates a consistently high yield of library from any amount of input DNA (including the PCR-free version), which is enough for library quantification   and sequencing on Illumina instruments.

Applications
The Collibri PS DNA Library Preparation Kit is recommended for:
Whole genome sequencing
• Resequencing
• De novo assembly

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Collibri PS DNA Library Prep Kit, store at -20°C
• Collibri DNA Library Cleanup Kit, store at 4°C
• Collibri DNA CD Indexes, store at -20°C
Description The product contains modules A39099096, A38584096
Format Kit
Sample Type DNA
Sequencing Type Genome and DNA Sequencing
For Use With (Equipment) HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NovaSeq 6000
For Use With (Application) Whole Genome Sequencing, Resequencing, De Novo Assembly
Libraries Fragment Library
No. of Reactions 96 Preps
Product Line Collibri
Product Type PS DNA Library Prep Core Kit
Quantity 96 preps
Shipping Condition Dry Ice
Workflow Step Library Generation
Show More Show Less
Are Collibri DNA Library Prep Core kits compatible with other commercially available indexed adaptors? I'm interested in performing higher throughput sequencing.

Collibri DNA Library Core kits are compatible with other commercially available set of adaptors such as Illumina TruSeq full-length adaptors, with eight base indexes.

Can indexed adaptors or other modules be purchased separately for the Collibri DNA Library Prep kits?

Indexed adapters and other modules in the kit can be purchased separately. Please refer to thermofisher.com/collibriDNA and contact us for more details.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.