Learn More
Eisco™ Collision Ball Apparatus
SCI_ED
Catalog No. S12906 Shop All Eisco Products
Catalog No. S12906 Supplier Eisco™ Supplier No. COL12
Add to Cart
Add to Cart
Demonstrates concept of momentum, conservation of energy, and Newton's third law.
  • Can be used on any level surface
  • V-shape track includes five steel balls
Specifications
Product Type Collision Ball Apparatus