Learn More
StatLab Collodion Coated Centrifuge Tube Prefills
SDP
Catalog No. 21033357
Change view
Click to view available options
Quantity:
15 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-033-357 15 mL
21-033-358 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-033-357 Supplier StatLab Supplier No. CYS3015C
Add to Cart
Add to Cart
Allow samples to be concentrated from fine needle aspirations (FNAs) and cellular material from effusions
  • Minute samples are concentrated into a pellet encased in the Collodion bag
  • First the sample is placed in the Collodion coated tube and centrifuged
  • The Collodion bag is then removed from the tube with the concentrated sample pellet at the bottom
  • The Collodion bag is trimmed and the encased pellet is ready for processing
Specifications
Quantity 15 mL
Product Type Centrifuge Tube