Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Colon adenocarcinoma tissue Tissue Slides (Adenocarcinoma)- Paraffin
SDP

Catalog No. NBP261407 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 Slides
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP261407 5 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP261407 Supplier Novus Biologicals™ Supplier No. NBP261407
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results.

  • Layout: 1X1
  • Diameter: 4
  • Thickness: Single Tissue Slides
  • Pos: A01
  • Species: Human
  • Age: 46
  • Sex: F
  • Organ: Colon
  • Tissue Condition: Adenocarcinoma
  • Tissue Status: Metastasis to liver
  • Protein State: Moderately Differentiated
  • Applications: HEStain, IHC, IHC-P
  • This product is for research use only and is not approved for use in humans or in clinical diagnosis.

Specifications

For Use With (Application) Immunohistochemistry-Paraffin
Quantity 5 Slides
Storage Requirements Store at 4°C under sterile conditions. Do not freeze.
Reconstitution These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 62°C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.