Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Colon Tissue Slides (Adenocarcinoma)- Paraffin
SDP

Catalog No. NBP230257 Shop All Novus Products
Change view
Click to view available options
Species:
Human
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
NBP230257 Human
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP230257 Supplier R&D Systems™ Supplier No. NBP230257
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems.

These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.

Specifications

Cross Reactivity Human
Type Colon Tissue Slides (Adenocarcinoma)
Regulatory Status This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Species Human
Tissue Condition Adenocarcinoma Moderately Differentiated
Product Line Novus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.