Learn More
United Scientific™ Colony Counter
SDPSCI_ED
Catalog No. S000857 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Product Type:
Colony Counter
Catalog No. Product Type
S000857 Colony Counter
1 options
Prop 65
Catalog No. S000857 Supplier United Scientific™ Supplier No. CCOUNTER
Add to Cart
Add to Cart
Prop 65
Colony Counter aids in counting microorganisms that are far too small to count with the naked eye.
Microorganisms are far too small to count with the naked eye, but they form visible colonies when they join together. When a Petri dish is placed on a lighted base, the grid pattern is illuminated. The 12 by 12 grid contains 1 x 1 cm squares. 24 of the squares are broken down into smaller increments of 9 squares, each 1/3 cm in size. To calculate an average colony count per square, the visible colonies are added together and divided by the total number of squares. Since the Petri dish is close to the light source, it is recommended to use an LED bulb to reduce the risk of heat damaging the sample. The apparatus has a standard bulb base.The power cord has an inline on/off switch. Overall dimensions are 9 x 9 x 7 in., 3.1 pounds. Required but not included is a light bulb. Includes activity guide.
  • Use of LED bulbs are recommended to protect sensitive samples
  • Large viewing area
Specifications
Product Type Colony Counter
Display Type None