Learn More
Cole-Parmer CC-200 Stuart Digital Colony Counter; 90 to 230 VAC, 50/60 Hz
Supplier:  Cole-Parmer 1421204/DEL
Encompass_Preferred
This colony counter registers a count each time you press a felt tip marker to the Petri dish. Cumulative count may be read on the digital display and is confirmed by an audible tone (can be turned on or off). Adjust the pressure required to measure a count — suitable to each user.Sub-stage illumination is provided by energy-saving, bright LEDs for glare-free observation. Enhance counting of translucent or difficult-to-see colonies with the supplied black background filter. Calculate the average colony count over multiple plates using the averaging facility. Count results and statistics, including standard deviation, can be sent directly to a printer or computer using the supplied USB cable. Order the 1.7x and 3x magnification adapters separately for easier viewing of small colonies.Please Note: Effective June 2022, the Stuart Shaking Water Bath Model number SC6PLUS has now been rebranded Cole-Parmer under Model number CC-200.
Catalog No. NC0767133
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.