Learn More
Cellular Engineering Technologies CHO-Expressed Macrophage colony-stimulating factor (CSF1). Vial of lyophilized power (50µg)
Supplier:  Cellular Engineering Technologies GR100250C
Encompass_Preferred
Human Colony Stimulating Factor 1 (CSF1), also known as macrophage colony-stimulating factor (M-CSF), is a secreted cytokine that causes hematopoietic stem cells to differentiate into macrophages or other related cell types. This growth factor is involved in the proliferation, differentiation, and survival of hematopoietic precursor cells, especially mononuclear phagocytes, such as macrophages and monocytes.CSF1 stimulates increased phagocytic and chemotactic activity of macrophages and monocytes. For hematopoietic progenitors, CSF1 interacts with membrane receptor CSR1R or M-CSF-R and modulates proliferation.CSF1 includes an Fc-fusion tag from human IgG1.Lyophilized powder.
Catalog No. NC2753131