Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ Color-Coded Food Hoe

Catalog No. 17227050 Shop All Remco Products
Change view
Click to view available options
Color Code:
Blue
Green
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Code
17-227-050 White
17-227-049 Red
17-227-051 Yellow
17-227-047 Green
17-227-048 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-227-050 Supplier Vikan™ Supplier No. 29005
Add to Cart
Add to Cart

Color-coded food hoe is a unique scraper designed for both push and pull applications.

  • Connects with any Euro-thread handle to move product on flat or curved surfaces efficiently
  • Color-coded to reduce cross-contamination and allergen cross-contact
  • Has the strength to move almost any type of product without breaking under pressure
  • Lightweight and longlasting
  • Flat and arched edges make it ideal for use on straight or curved surfaces

Specifications

Product Type Food Hoe
Color Code White
Color-coded Yes
Material Nylon
Size Large
Certifications/Compliance FDA Compliant
Dimensions (L x W) 11.25 x 2 in.
For Use With (Application) Moving products
No. per Pack 1 Ea.
Weight (English) 1.13 lbs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.