Learn More
Specifications
Specifications
| Adhesive Type | Self-adhesive |
| Color | Dark Green |
| Format | Roll |
| Material | Paper |
| Product Type | Color-coded Label |
| For Use With (Application) | General and industrial identification |
| Quantity | 500/Sheet |
| Shape | Round |
| Diameter (English) | 1.5 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.