Learn More
Thermo Scientific™ Color-coded Sample Alert Labels
GREENER_CHOICE
Catalog No. 05719534
Change view
Click to view available options
Color:
Blue
Pink
Red
Yellow
Product Type:
Sample Label, Hydrochloric Acid
Sample Label, Nitric Acid
Sample Label, Short Holding Time
Sample Label, Sodium Hydroxide
Sample Label, Sulfuric Acid
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type
05-719-534 Blue Sample Label, Hydrochloric Acid
05-719-532 Red Sample Label, Nitric Acid
05-719-531 Yellow Sample Label, Sulfuric Acid
05-719-535 Pink Sample Label, Sodium Hydroxide
05-719-537 Red Sample Label, Short Holding Time
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 05-719-534 Supplier Thermo Scientific™ Supplier No. 5030007
Add to Cart
Add to Cart
Increase the safety and efficiency of sample handling and identification with color-coded sample alert labels.
Increase the safety and efficiency of sample handling and identification with Thermo Scientific™ Color-Coded Sample Alert Labels. Each label is color-coded with the chemical preservative type for quick reference and safe handling. Sticks securely to plastic or glass sample containers.
  • Color-coded adhesive “dot“ with chemical preservative or short holding time alert
  • Roll of 1000 labels
  • Provides easy sample aliquot identification and enhances safe handling information visibility
  • Sticks securely to plastic or glass
TRUSTED_SUSTAINABILITY
Specifications
Certification Parameters None
Color Blue
No. per Case 1,000
Certifications/Compliance None
Height (Metric) 25.4 mm
Product Type Sample Label, Hydrochloric Acid
Height (English) 1 in. (round)