Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Color Paddles
SCI_ED

Catalog No. S8834
Change view
Click to view available options
Quantity:
3 sets of six
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S8834 3 sets of six
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S8834 Supplier Learning Resources, Inc Supplier No. LER 0352
Add to Cart
Add to Cart

Demonstrate color-mixing principles.

  • Transparent 6 in. (15.2cm) paddles
  • Six colors with three textures

Order Info

Sold in three sets of six.

Specifications

Product Type Color Paddle Set
For Use With (Application) To demonstrate the principles of color mixing
Quantity 3 sets of six
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.