Learn More
Color Reagent for Chloride, Certified, LabChem™
Specifications
Specifications
| Chemical Name or Material | Color Reagent for Chloride |
| Color | Orange |
| CAS Max % | 81.605, 14.937, 3.03, 0.315, 0.063 |
| CAS | 67-56-1, 7782-61-8, 7697-37-2, 592-85-8 |
| Linear Formula | Fe(NO3)3·9H2O |
| Solubility Information | Soluble in water |
| Formula Weight | 404.02 |
| Grade | Certified |
| Identification | Passes Test |
| Packaging | Amber Glass |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.