Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Color Reagent for Chloride, Certified, LabChem™

Catalog No. LC132602
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC132602 1 L
LC132601 500 mL
LC132604 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC132602 Supplier LabChem, Inc. Supplier No. LC132602
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Color Reagent for Chloride
Color Orange
CAS Max % 81.605, 14.937, 3.03, 0.315, 0.063
CAS 67-56-1, 7782-61-8, 7697-37-2, 592-85-8
Linear Formula Fe(NO3)3·9H2O
Solubility Information Soluble in water
Formula Weight 404.02
Grade Certified
Identification Passes Test
Packaging Amber Glass
Suitability Passes Test
Decomposition Information Mercury; Carbon monoxide; Carbon dioxide; Sulfur compounds; Nitrogen oxides
Quantity 1 L
Physical Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.