Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Color Reagent, for Nitrate Determination, Ricca Chemical
SDP

Catalog No. 22331 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22331 4 L
2233-4 120 mL
2233-16 500 mL
223332 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22331 Supplier Ricca Chemical Company Supplier No. 22331
Add to Cart
Add to Cart

For Nitrate Determination

Specifications

Chemical Name or Material Color Reagent
Color Colorless
CAS 7732-18-5, 7664-38-2, 63-74-1, 1465-25-4
CAS Min % 81.0, 13.68, 3.24, 0.16
CAS Max % 84.0, 14.23, 3.37, 0.17
Grade Laboratory
Packaging Amber Glass Bottle
pH Acidic
Quantity 4 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.