Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Color Reagent, for Nitrite Determination, Ricca Chemical
SDP

Catalog No. 2233516 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22335-16 500 mL
2233-532 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22335-16 Supplier Ricca Chemical Company Supplier No. 2233.516
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sulfanilamide/N-(1-Naphthyl)ethylenediamine Dihydrochloride Solution

Specifications

Chemical Name or Material Color Reagent
Color Colorless
CAS 1465-25-4, 63-74-1, 7664-38-2, 7732-18-5
Grade Laboratory
Packaging Amber Glass Bottle
pH Acidic
Quantity 500 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.