Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Color Reagent for Nitrogen (Nitrate), For Automated Cadmium Reduction Method, LabChem™

Catalog No. LC132801
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC132801 500 mL
LC132802 1 L
LC132804 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC132801 Supplier LabChem, Inc. Supplier No. LC132801
Add to Cart
Add to Cart

Specifications

Color Clear, slight pink to tan
Format Liquid
Packaging Amber Glass
Quantity 500 mL
Storage Requirements Refrigerate
Type Color Reagent for Nitrogen (Nitrate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.