Learn More
Color Reagent for Nitrogen (Nitrate), For Automated Cadmium Reduction Method, LabChem™
Specifications
Specifications
| Color | Clear, slight pink to tan |
| Format | Liquid |
| Packaging | Amber Glass |
| Quantity | 1 L |
| Storage Requirements | Refrigerate |
| Type | Color Reagent for Nitrogen (Nitrate) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.