Learn More
Sigma Aldrich Fine Chemicals Biosciences Color Reference Solutions | Purity: N/A | 20 x 10 mL Vials | Brown, Yellow, and Red Hue Standards for Pharmaceutical Quality Control Testing per USP Guidelines
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 875741SETF
Encompass
Color Reference Solutions USP | Purity: N/A | M. W.: N/A | CAS No.: N/A | MDL No.: N/A | 20 x 10mL vials
These analytical standard solutions exhibit a range of color intensities that cover brown, yellow, and red hues. The color reference standard solutions meet specifications outlined in the United States Pharmacopeia and are intended for use in the pharmaceutical industry to calibrate instrumentation and control the degree of solution coloration during quality control testing procedures. The set of 20 x 10mL vials covers the color range of brown, yellow, and red hues and is used to calibrate and control solution coloration for pharmaceutical quality control testing according to USP color reference standards.
Catalog No. NC1962518
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.