Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Color Reference Solutions acc. to Ph Eur

Catalog No. 01184196 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Color Reference; Grade: Analytical Standard
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-184-196 Color Reference; Grade: Analytical Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-184-196 Supplier MilliporeSigma™ Supelco™ Supplier No. 839521SETF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Color Reference Solutions - Analytical standard, Set of colors B, BY, Y, GY, R.

  • The kit contains set of brown(83951), yellow (83883), red (87448), greenish-yellow (82995), and brownish-yellow (86293) color reference standards. The solutions are produced from three primary metal salt solutions of ferric(III) chloride, cobalt(II) chloride, and copper(II) sulfate pentahydrate, in accordance with chapter 2.2.2 'Degree of coloration of liquids' of the European Pharmacopoeia (Ph. Eur.).
  • Application
    • They are used as calibration and/or quality control standards to measure the degree of coloration of liquids in the brown-yellow-red color range.
  • Features and Benefits
    • Ready-to-use solutions
    • Prepared and qualified in accordance with Ph. Eur.
    • Available in tamper-evident 2 mL ampules
    • Expiry dated on the label with a limited shelf life
    • Accompanied with a detailed certificate of analysis for an effective quality management system
  • Components
  • This is a set of the following color reference solutions sets according to PhEur:
    • 83951-1SET-FColor Reference Solutions B acc. to PhEur - 10 x 2 mL ampules
    • 86293-1SET-FColor Reference Solutions BY acc. to PhEur - 8 x 2 mL ampules
    • 83883-1SET-FColor Reference Solutions Y acc. to PhEur - 8 x 2 mL ampules
    • 82995-1SET-FColor Reference Solutions GY acc. to PhEur - 8 x 2 mL ampules
    • 87448-1SET-FColor Reference Solutions R acc. to PhEur - 8 x 2 mL ampules
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule of 2 mL
Certifications/Compliance European Pharmacopoeia
Type Color Reference Standard
Industry Type Food and Beverage, Flavors and Fragrances, Glass and Ceramics, Pharmaceutical, Coatings
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.