Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Color Standard, APHA/Hazen (Platinum-Cobalt), Color = 30, Ricca Chemical
SDP

Catalog No. R2230030100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R2230030100 100 mL
2230030-16 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R2230030100 Supplier Ricca Chemical Company Supplier No. 2230.030100
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Color Standard
Packaging Amber Glass Bottle
Name Note Color = 30
CAS Min % 97.0, 0.6, 0.01, 0.01
CAS Max % 100.0, 0.62, 0.01, 0.01
CAS 7647-01-0, 16921-30-5, 7791-13-1, 7647-01-0, 7732-18-5
Methods APHA / Hazen (Platinum-Cobalt)
Physical Form Liquid
Color Brown/Yellow to Yellow
Boiling Point 100°C
Quantity 100 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.