Learn More
Color Standard, APHA/Hazen (Platinum-Cobalt), Color = 400, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Chemical Name or Material | Color Standard |
| Packaging | Amber Glass Bottle |
| Name Note | Color = 400 |
| CAS Min % | 93.0, 4.64, 0.05, 0.04 |
| CAS Max % | 97.0, 4.83, 0.05, 0.04 |
| CAS | 7647-01-0, 16921-30-5, 7791-13-1, 7647-01-0, 7732-18-5 |
| Methods | APHA / Hazen (Platinum-Cobalt) |
| Physical Form | Liquid |
| Color | Brown/Yellow to Yellow |
| Boiling Point | 100°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.