Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Color Standard, APHA/Hazen (Platinum-Cobalt), Color = 5, Ricca Chemical
SDP

Catalog No. R2230005250 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R2230005250 250 mL
R2230005100 100 mL
2230005-16 500 mL
R22300054C 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R2230005250 Supplier Ricca Chemical Company Supplier No. 2230.0058
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Color Standard, APHA/Hazen (Platinum-Cobalt), Color=5
Packaging Amber Glass Bottle
CAS Min % 99.95, 0.04, 0.0013, 0.001
CAS Max % 99.95, 0.04, 0.0013, 0.001
CAS 16921-30-5, 7791-13-1, 7647-01-0, 7732-18-5
Density 1.001
Physical Form Liquid
Color Brown/Yellow
pH <1
Melting Point 0°C
Boiling Point 100°C
Solubility Miscible with water
Quantity 250 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.