Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Color Standard, APHA/Hazen (Platinum-Cobalt), Color = 500, Ricca Chemical
SDP

Catalog No. 22304 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
200 mL
250 mL
500 mL
1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22304 120 mL
R2230000200 200 mL
R2230000250 250 mL
2230-16 500 mL
2230-32 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22304 Supplier Ricca Chemical Company Supplier No. 22304
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Specifications

Chemical Name or Material Color Standard
Packaging Amber Glass Bottle
Name Note Color= 500
CAS Min % 87.0, 10.32, 0.11, 0.09
CAS Max % 91.0, 10.74, 0.12, 0.09
CAS 16921-30-5, 7791-13-1, 7647-01-0, 7732-18-5
Melting Point Approximately 0°C
Quantity 120 mL
Grade APHA/Hazen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.