Learn More
Color Standard, Pt-Co, 100 Units APHA, Certified, LabChem™
Specifications
Specifications
| Chemical Name or Material | Color Standard |
| Name Note | Pt-Co, 100 Units APHA |
| Packaging | Amber Glass |
| CAS Max % | 99.08, 0.87, 0.03, 0.02 |
| CAS | 16921-30-5, 7791-13-1, 7647-01-0, 7732-18-5 |
| Identification | Passes Test |
| Density | 1g/mL |
| Physical Form | Liquid |
| Linear Formula | K2(PtCl6) |
| Color | Orange |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.