Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Color Standard, Pt-Co, 500 Units APHA, Stock Solution, Certified, LabChem™

Catalog No. LC133001
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC133001 500 mL
LC133007 125 mL
LC133002 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. LC133001 Supplier LabChem, Inc. Supplier No. LC133001
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Specifications

Chemical Name or Material Color Standard
Name Note Pt-Co, 500 Units APHA, Stock Solution
Packaging Amber Glass
CAS Max % 95.43, 4.34, 0.13, 0.1
CAS 16921-30-5, 7791-13-1, 7647-01-0, 7732-18-5
Identification Passes Test
Physical Form Liquid
Linear Formula K2(PtCl6)
UN Number UN1789
Color Orange
Solubility Soluble in water
Quantity 500 mL
Formula Weight 485.99
Grade Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.