Learn More
Description
- Color Staph latex reagent reacts with characteristics
- Positive reaction: aggregation of smooth latex suspension, red agglutination with blue background
- Detects MRSA
- Includes liquid controls
- Sensitivity: > 99%
- Specificity: > 99%
- Certifications:
- CPT code 87449
- 510(k)- K830783
Specifications
Specifications
| Certifications/Compliance | CPT code 87449, 510(k)- K830783 |
| Detectable Analytes | MRSA |
| Sensitivity | >99% |
| Type | Color Staph Kit |
| Includes | Liquid Controls |
| Quantity | 50 Tests |
| Specificity | > 99% |
| Test Time | 10 to 30 sec. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
