Learn More
Description
- Made of durable polyester
- Available in five colors
- Choose from two sizes: 3.5 x 2.5 in. or 5 x 3.5 in.
Specifications
Specifications
| Product Type | Laminating Pouch |
| Color | Yellow |
| For Use With (Equipment) | Brady Label Printers |
| Material | Polyester |
| Dimensions (L x W) | 3.5 x 2.5 in. (W x H) |
| Description | Holes Not Included |
| Shape | Rectangle |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.