Learn More
Profoldin Colorimetric DTT Assay Kit
Supplier:  Profoldin DTT100C
Encompass
The Colorimetric DTT Assay Kit (Catalog No. DTT100C) measures light absorbance at 590 nm. It is based on the strong reducing property of DTT. The assay is not interfered by phosphate buffer, mono thio compounds such as cysteine or chelators. It is interfered by NADH or NADPH.
Catalog No. NC1836973
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.