Learn More
Epredia™ Colormark™ Slide Cartridges forSHUR/Mark™ Labeling Systems
Catalog No. 22035814 Shop All Epredia Products
Change view
Click to view available options
Color:
Blue
Lilac/Black
Pink
White
Yellow
Coating:
Adhesion
Non-adhesion
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Coating
22-035814 White Non-adhesion
22-035815 Blue Non-adhesion
22-038325 Blue Adhesion
22-038328 Lilac/Black Adhesion
22-035817 Pink Non-adhesion
22-038323 Pink Adhesion
22-038324 White Adhesion
22-035818 Yellow Non-adhesion
22-038327 Yellow Adhesion
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 22-035814 Supplier Epredia™ Supplier No. CM4951W001
Add to Cart
Add to Cart
This item is not returnable. View return policy
Background color makes etched patient information more visible
  • Brightly coated with contrasting background color
  • Self-contained packages ready for loading into your SHUR/Mark slide labeling system
  • Simply remove the cover from the slide package and drop the sleeve into the loading bin of the SHUR/Mark labeling system
  • Available with Plus formula to provide the utmost adhesion
Specifications
Color White
Coating Non-adhesion
Quantity 144 Pk., 10 Pk./Cs.