Learn More
Cytiva COLUMN CLAMP O.D. 10?21 MM
Supplier:  Cytiva 28956319
Encompass
Column clamp for use with KTA systems, such as KTA pure, KTA avant and KTA start. Fits columns with OD between 10 and 21 mm. Pack of five.0
Catalog No. 50-105-5664