Description
- Economical protection for your guard or analytical HPLC column
- No wrenches or tools required to install, fingertight to 5,000 psi
- Lower dead volume than conventional filters with holders
- Universal connection is compatible with all manufacturers fittings
The PEEK filter body contains a HiFlo filter element and is designed for maximum filtration of particulate matter with mimimal dead volume or backpressure. As soon as an increase in backpressure is observed, simply remove and dispose of the Supelco columnSaver and install a new one. The direct connect design is compatible with all 1/16 in.,10-32 internal fitting ports regardless of the manufacturer.
Specifications
Specifications
| Size (Metric) | 2.0 μm |
| Description | 10/Pk. |