Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Com-Pac International™ SAF-T-ZIP™ Specimen Bags
Cost-effective alternative for specimen collection
$202.00
Specifications
Closure Type | Zip Seal |
---|---|
Dimensions (L x W) | 6 x 9 in. (15.2 x 22.9cm) |
Length (English) | 6 in. |
Length (Metric) | 15.2 cm |
Width (English) | 9 in. |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
22-043-228
|
Com-Pac International™
7210GR REV B |
Green |
Case of 1000 for $202.00
|
|
|||||
22-043-225
|
Com-Pac International™
7210RD REV B |
Red |
Case of 1000 for $202.00
|
|
|||||
Description
- Color-coded zippers for easy sorting
- Convenient document pouch separates documents from the specimen
- Printed with the OSHA-required biohazard symbol
- Measure 6 × 9 in. (15.2 × 22.9cm)
Specifications
Zip Seal | |
6 in. | |
9 in. |
6 x 9 in. (15.2 x 22.9cm) | |
15.2 cm | |
22.9 cm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title