Learn More
Cole-Parmer Food Waterproof Thermocouple Thermometer, Type-K and T with Single Input
Supplier:  Cole-Parmer 9002533
Encompass_Preferred
COMARK N9094 Food Waterproof Thermocouple Thermometer, Type-K and T with Single Input, Waterproof design - ideal for working in adverse conditionsIP68-rated case is water- and dustproofExceptional battery life of up to 10 yearsAccepts both thermocouple and thermistor probes
Catalog No. NC1573925
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.