Learn More
Liofilchem™ ComASP Ceftolozane-tazobactam
SDP
Catalog No. 22778090 Shop All Liofilchem Products
Change view
Click to view available options
Quantity:
4 Systems (panels)
Catalog No. Quantity
22-778-090 4 Systems (panels)
1 options
Non-hazardous according to current legislation but fall within the specific field of application where a safety data sheet must be supplied because they can cause phenomena of sensitisation in sensitive subjects if they come into contact with the skin.
Catalog No. 22-778-090 Supplier Liofilchem™ Supplier No. 75006
Only null left
Add to Cart
Add to Cart
Non-hazardous according to current legislation but fall within the specific field of application where a safety data sheet must be supplied because they can cause phenomena of sensitisation in sensitive subjects if they come into contact with the skin.
The system is used to perform the broth microdilution (BMD) method for the antimicrobial susceptibility testing of ceftolozane-tazobactam as recommended by international standards (i.e. CLSI, EUCAST, ISO) but in a simpler and less time-consuming way
  • 4 Systems (panels) of ComASP C/T (panels individually packed in foil with silica gel desiccant
  • 8 Tubes of Mueller Hinton II Broth (3.6 mL)
  • Sealing Film
  • Instructions Sheet and Test Results Form
Specifications
Antimicrobial Agents Ceftolozane-tazobactam
Concentration 0.008 to 128 μg/mL
Content And Storage Store ComASP™ C/T at 2°C to 8°C in the original packaging. Once an envelope is opened the panel should be used within 7 days. Keep away from direct sunlight and direct heat
Product Type Panel
Sterility Non-sterile
Test Type Media Testing
Quantity 4 Systems (panels)