Learn More
Liofilchem™ ComASP Vancomycin
SDP
Catalog No. 22778091 Shop All Liofilchem Products
Change view
Click to view available options
Quantity:
4 Systems (panels)
Catalog No. Quantity
22-778-091 4 Systems (panels)
1 options
Non-hazardous according to current legislation but fall within the specific field of application where a safety data sheet must be supplied because they can cause phenomena of sensitisation in sensitive subjects if they come into contact with the skin.
Catalog No. 22-778-091 Supplier Liofilchem™ Supplier No. 75007
Add to Cart
Add to Cart
Non-hazardous according to current legislation but fall within the specific field of application where a safety data sheet must be supplied because they can cause phenomena of sensitisation in sensitive subjects if they come into contact with the skin.
The system is used to perform the broth microdilution (BMD) method for the antimicrobial susceptibility testing of vancomcyin as recommended by international standards (i.e. CLSI, EUCAST, ISO) but in a simpler and less timeconsuming way.
  • 4 Systems (panels) of ComASP Vancomycin (panels individually packed in foil with silica gel desiccant)
  • 8 Tubes of Mueller Hinton II Broth (3.6 mL)
  • 15 Sealing Films
Specifications
Antimicrobial Agents Vancomycin
Concentration 0.008 to 128 μg/mL
Content And Storage Store ComASP™ Vancomycin at 2°C to 8°C in the original packaging. Once an envelope is opened the panel should be used within 7 days. Keep away from direct sunlight and direct heat.
Product Type Panel
Sterility Non-sterile
Test Type Media Testing
Quantity 4 Systems (panels)