Description
- Offset connector allows the user to rotate the cartridge into 3 different positions improving visibility
- Saf-Twist design quickly attaches the cartridge to any Dentec Safety half-mask or full face air-purifying respirator Facepiece.
Specifications
Specifications
| Filter Type | Combination Ammonia/Methylamine and P100 |
| Material | Plastic |
| Product Type | Respirator Accessories |
| For Use With (Equipment) | Masks |
| Includes | Cartridges and filters |
| Quantity | 60/Box |