Learn More
Ampco Safety Tools Combination Wrench
GREENER_CHOICESDP
Catalog No. 19173374 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Size:
6 mm
7 mm
20 mm
23 mm
25 mm
26 mm
29 mm
55 mm
70 mm
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-173-374 26 mm
19-173-354 6 mm
19-173-355 7 mm
19-173-368 20 mm
19-173-371 23 mm
19-173-373 25 mm
19-173-377 29 mm
19-173-386 55 mm
19-173-389 70 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog No. 19-173-374 Supplier Ampco Safety Tools Supplier No. 1336
Add to Cart
Add to Cart
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
AMPCO™ Combination Wrench performs in high-risk and demanding environments.
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Material Aluminum Bronze
Size 26 mm
Certifications/Compliance Factory Mutual (FM) Approved
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.