Learn More
Med Vet International Abdominal Abd Combine Dressing, 5-in X 9-in, Non-Sterile, Dukal, 400/Case
Supplier:  Med Vet International 5940
Encompass_Preferred
Combine pads consist of three key layers: outer non-woven layer to wick, fluff filler to absorb and disperse fluid and wet proof moisture barrier to retard fluid strike-through. These pads act as a reservoir, reduces pooling and protects against contamination. Non-sterile, 400/Case
Catalog No. 50-283-2230
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.