Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Combitips™ Plus Pipet Tips

Catalog No. 21381327 Shop All Eppendorf North America Products
Change view
Click to view available options
Nonsterile; Volume: 0.5mL; Dispensing range: 5 to 100μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
21-381-327 Nonsterile; Volume: 0.5mL; Dispensing range: 5 to 100μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-381-327 Supplier Eppendorf™ Supplier No. 022266101

missing translation for 'validMainframeMsg'

Positive displacement for correct volumes

Tips function according to the positive-displacement principle, ensuring that the correct volume is dispensed with regard to the density and viscosity of the liquid used. The positive displacement system provides safer dispensing of radioactive and toxic substances as well as solutions with high vapor pressure. No aerosols are formed.

  • For use with Eppendorf Repeaters™ (original, Plus, Pro, stream and Xstream); and EDOS (automatic dispensing system)
  • Available in nine volumes from 0.1 to 50mL
  • Reliable results ensured by positive-displacement system
  • Fine tip of Combitip provides exact dispensing without additional pipet tip
  • Polyethylene plunger and polypropylene cylinder make these tips ideal for use with aggressive reagents
  • Choose from sterile and nonsterile Combitips Plus Tips and Eppendorf Biopur™ tips, which are certified sterile as well as pyrogen-, RNase-, DNA-, and ATP-free

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.5 mL
Sterility Non-sterile
Tip Style Specialty, Positive Displacement
For Use With (Equipment) Eppendorf Repeater, EDOS (Automatic Dispensing System)
Product Line CombiTips Plus
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.