Learn More
Grainger Combination Wrench, 13mm, 6in OAL
Supplier:  Grainger 21A295
Encompass_Preferred
The Grainger 13mm Combination Wrench is a 6 inch overall length chrome vanadium steel wrench with a 15 degree offset head. It has a satin chrome finish to resist corrosion. The wrench has a limited lifetime warranty and is made in Taiwan. This wrench can be used to loosen or tighten 13mm nuts and bolts in tight spaces.
Catalog No. NC2603808
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.