Learn More
Description
Specifications
Specifications
| Content And Storage | The product requires storage at both 2 - 8°C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date. |
| Format | Kit |
| For Use With (Application) | A simple and effective method for evaluating DNA damage in cells |
| Includes | ES unit |
| Product Type | Comet Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
