Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CometAssay Electrophoresis System
SDP

Catalog No. 4250050ES Shop All Novus Products
Change view
Click to view available options
Content And Storage:
The product requires storage at both 2 - 8°C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
42-500-50ES The product requires storage at both 2 - 8°C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Caution! Electrical Hazard! This equipment is designed for use with a DC power supply providing up to 250 VDC. Careless handling can result in electrical shock. To avoid any risk of injury, the instrument should be operated in accordance with the instruction provided.
Catalog No. 42-500-50ES Supplier R&D Systems™ Supplier No. 4250050ES
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Caution! Electrical Hazard! This equipment is designed for use with a DC power supply providing up to 250 VDC. Careless handling can result in electrical shock. To avoid any risk of injury, the instrument should be operated in accordance with the instruction provided.

Electrophoresis System for the CometAssay™

  • Overcomes assay variability first by placing an acrylic overlay on top of an elevated slide tray to maintain optimal buffer height for DNA migration
  • Secondly, a constant buffer temperature is maintained using an external chamber under the tank to cool the glass slide platform and buffer chamber
  • Electrophoresis time for optimal DNA migration is achieved in 30 minutes with Alkaline (pH>13) Electrophoresis Solution, or 45 minutes with Neutral Electrophoresis Buffer at 1V/cm
  • The smoke grey colored unit is designed to minimize exposure to UV light but still allow visual inspection
  • Third, specially designed slide trays are also provided to accommodate 2, 20 and 96 well slides and maintain proper slide orientation during electrophoresis
  • CometAssay™ Electrophoresis System II, CometAssay™ Alkaline Control Cells, and CometAssay™ Neutral Control Cells enable investigators to consistently optimize their comet assays for highly reproducible results, and to standardize electrophoresis methods between individual users and laboratories

Specifications

Content And Storage The product requires storage at both 2 - 8°C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
For Use With (Application) A simple and effective method for evaluating DNA damage in cells
Includes Electrophoresis Tank with Leveling Bubble, Safety Lid & Cables, 2/20 Well Slide Trays, and Slide Tray Overlay
Product Line R&D Systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.