Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CometAssay LMAgarose
SDP

Catalog No. 425050002 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-505-0002 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 42-505-0002 Supplier R&D Systems™ Supplier No. 425050002

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

1 bottle (100 mL) of a CometAssay™ LMAgarose

  • Store bottle at 2-8 °C
  • CometAssay™ LMAgarose is ready to use once molten
  • Loosen the cap to allow for expansion then heat the bottle in a 90-100 °C water bath for 5 minutes, or until the agarose is molten (microwaving is not recommended)
  • Place the bottle in a 37 °C water bath for at least 20 minutes to cool
  • The LMAgarose will remain molten at 37 °C for sample preparation indefinitely
  • CometAssay™ LMAgarose is 1% low melting agarose in PBS
  • Designed for use with CometAssay™, FLARE™, and CometChip™ Kits

Specifications

For Use With (Application) CometAssay™, FLARE™, and CometChip™ Kits
Quantity 100 mL
Storage Requirements Store the unopened product at 2 - 8°C. Do not use past expiration date.
Product Line R&D Systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.